Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318915.1 | NP_001305844.1 | 9 | 4443 | 65 | |
NM_002338.4 | NP_002329.2 | 7 | 4374 | 65 | ||
GenBank mRNA | AK094535.1 | - | 2 | 2063 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |