Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000371408.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000427197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000483386.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000494992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000603665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
ENST00000605679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_001134658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008492.hg.1 |
NM_153226.3 |
Refseq |