Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150496.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150496.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150496.hg.1 |
TC1800007360.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150496.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150496.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150495.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150495.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150495.hg.1 |
TC1800007360.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150495.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150495.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150494.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150494.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150494.hg.1 |
TC1800007360.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150494.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150494.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000262094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000586594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
ENST00000592334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007360.hg.1 |
NM_004163.4 |
Refseq |