Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090434.1 | - | 3 | 5281 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |