Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006716896.2 | XP_006716959.1 | 1 | 113 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |