Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256799.2 | NP_001243728.1 | 7 | 807 | 139 | |
NM_001289745.1 | NP_001276674.1 | 8 | 865 | 139 | ||
NM_001289746.1 | NP_001276675.1 | 7 | 759 | 139 | ||
NM_002046.5 | NP_002037.2 | 8 | 773 | 139 | ||
GenBank mRNA | AB062273.1 | - | 8 | 637 | 139 | |
AF261085.1 | - | 8 | 688 | 139 | ||
AK026525.1 | - | 8 | 710 | 139 | ||
AK308198.1 | - | 8 | 661 | 139 | ||
AY007133.1 | - | 8 | 637 | 139 | ||
AY633612.1 | - | 7 | 609 | 139 | ||
BC001601.1 | - | 8 | 639 | 3543589 | 139 | |
BC004109.2 | - | 8 | 637 | 3678259 | 139 | |
BC009081.1 | - | 8 | 646 | 3869809 | 139 | |
BC013310.2 | - | 8 | 645 | 3538383 | 139 | |
BC020308.1 | - | 7 | 623 | 139 | ||
BC023632.2 | - | 7 | 609 | 4842418 | 139 | |
BC025925.1 | - | 8 | 642 | 2989348 | 139 | |
BC026907.1 | - | 7 | 623 | 4454083 | 139 | |
BC029340.1 | - | 8 | 631 | 139 | ||
BC029618.1 | - | 8 | 639 | 3876535 | 139 | |
BC083511.1 | - | 8 | 637 | 3536969 | 139 | |
BT006893.1 | - | 7 | 585 | 139 | ||
CR407671.1 | - | 7 | 585 | 139 | ||
DQ403057.1 | - | 6 | 510 | 139 | ||
EF036498.1 | - | 4 | 323 | 139 | ||
EU668321.1 | - | 8 | 687 | 139 | ||
J02642.1 | - | 7 | 586 | 139 | ||
M17851.1 | - | 7 | 618 | 139 | ||
M33197.1 | - | 8 | 645 | 139 | ||
X01677.1 | - | 7 | 645 | 139 | ||
X53778.1 | - | 8 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Amplicon spans exons and probe does not span exons |