Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003540.3 | NP_003531.1 | 1 | 305 | 114 | |
GenBank mRNA | BC069288.1 | - | 1 | 305 | 7262669 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625426 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813707 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15793761 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813689 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625441 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625444 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813686 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813697 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813702 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625434 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813691 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813683 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813684 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15793756 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15793760 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P359540 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P359540 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 25904 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56239 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P31701835 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P359540 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P359540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625444 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625450 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625449 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813711 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813689 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813689 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625444 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625449 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625450 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813711 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813683 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P359540 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625448 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625442 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625445 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625444 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625446 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04625440 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04625441 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P359540 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 10378 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P31701833 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625409 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625399 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625421 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793752 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625397 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625388 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625370 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625449 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625431 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793755 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625471 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625382 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625451 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625439 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625425 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625404 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625446 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625417 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625448 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625413 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625490 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625487 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625369 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625407 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625391 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625375 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625480 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625453 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625472 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625455 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625411 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625460 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625435 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625491 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625456 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625429 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625478 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625442 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625475 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625469 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625402 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625444 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625372 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625485 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625427 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625483 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_003540_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117425 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117423 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117424 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8117422 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208026_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738103_at | - | - |
Affymetrix Human Genome U219 Array | 11738103_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06001768.hg.1 | TC06000175.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06001768.hg.1 | NM_003540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149373.hg.1 | NM_003540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149373.hg.1 | NM_003540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149373.hg.1 | TC0600007289.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 206951_at | - | - |
Human Genome U133 Plus 2.0 Array | 206951_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 234960_at | - | - |
Human Genome U133 Plus 2.0 Array | 208580_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 214634_at | - | - |
Human Genome U133 Plus 2.0 Array | 208076_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 234960_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208181_at | - | - |
Human Genome U133 Plus 2.0 Array | 232035_at | - | - |
Human Genome U133 Plus 2.0 Array | 207046_at | - | - |
Human Genome U133 Plus 2.0 Array | 208181_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214634_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214516_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214516_at | - | - |
Human Genome U133 Plus 2.0 Array | 208026_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 207046_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 232035_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 205967_at | - | - |
Human Genome U133 Plus 2.0 Array | 208180_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 208580_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208180_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 208026_at | - | - |
Human Genome U133 Plus 2.0 Array | 208076_at | - | - |
Human Genome U133 Plus 2.0 Array | 208046_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208046_at | - | - |
Human Genome U133 Plus 2.0 Array | 205967_PM_at | - | - |
Human Genome U133A 2.0 Array | 214634_PM_at | - | - |
Human Genome U133A 2.0 Array | 208046_PM_at | - | - |
Human Genome U133A 2.0 Array | 208046_at | - | - |
Human Genome U133A 2.0 Array | 208076_PM_at | - | - |
Human Genome U133A 2.0 Array | 208181_PM_at | - | - |
Human Genome U133A 2.0 Array | 207046_at | - | - |
Human Genome U133A 2.0 Array | 208181_at | - | - |
Human Genome U133A 2.0 Array | 206951_at | - | - |
Human Genome U133A 2.0 Array | 208180_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 208580_x_at | - | - |
Human Genome U133A 2.0 Array | 214516_at | - | - |
Human Genome U133A 2.0 Array | 214516_PM_at | - | - |
Human Genome U133A 2.0 Array | 208026_at | - | - |
Human Genome U133A 2.0 Array | 205967_at | - | - |
Human Genome U133A 2.0 Array | 205967_PM_at | - | - |
Human Genome U133A 2.0 Array | 206951_PM_at | - | - |
Human Genome U133A 2.0 Array | 207046_PM_at | - | - |
Human Genome U133A 2.0 Array | 208076_at | - | - |
Human Genome U133A 2.0 Array | 208180_s_at | - | - |
Human Genome U133A 2.0 Array | 208580_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 208026_PM_at | - | - |
Human Genome U133A 2.0 Array | 214634_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003540_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw588937 | - | - |
Human Genome U95 Set | 34027_f_at | - | - |
Human Genome U95 Set | 33049_PM_at | - | - |
Human Genome U95 Set | 31521_f_at | - | - |
Human Genome U95 Set | 152_PM_f_at | - | - |
Human Genome U95 Set | 762_f_at | - | - |
Human Genome U95 Set | 152_f_at | - | - |
Human Genome U95 Set | 33049_at | - | - |
Human Genome U95 Set | 762_PM_f_at | - | - |
Human Genome U95 Set | 34027_PM_f_at | - | - |
Human Genome U95 Set | 31521_PM_f_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208026_at2 | - | - |
Human Genome U95 Set | 59154_PM_at | - | - |
Human Genome U95 Set | 59154_at | - | - |
Human Genome U95 Set | 57785_at | - | - |
Human Genome U95 Set | 57785_PM_at | - | - |
Human Genome U95 Set | 81125_PM_at | - | - |
Human Genome U95 Set | 81125_at | - | - |
Human Genome U133 Set | 214516_at | - | - |
Human Genome U133 Set | 208046_PM_at | - | - |
Human Genome U133 Set | 208180_s_at | - | - |
Human Genome U133 Set | 207046_at | - | - |
Human Genome U133 Set | 208026_at | - | - |
Human Genome U133 Set | 214516_PM_at | - | - |
Human Genome U133 Set | 208580_PM_x_at | - | - |
Human Genome U133 Set | 208180_PM_s_at | - | - |
Human Genome U133 Set | 214634_PM_at | - | - |
Human Genome U133 Set | 207046_PM_at | - | - |
Human Genome U133 Set | 208181_PM_at | - | - |
Human Genome U133 Set | 208181_at | - | - |
Human Genome U133 Set | 205967_PM_at | - | - |
Human Genome U133 Set | 214634_at | - | - |
Human Genome U133 Set | 208076_at | - | - |
Human Genome U133 Set | 208026_PM_at | - | - |
Human Genome U133 Set | 208046_at | - | - |
Human Genome U133 Set | 206951_PM_at | - | - |
Human Genome U133 Set | 208076_PM_at | - | - |
Human Genome U133 Set | 205967_at | - | - |
Human Genome U133 Set | 206951_at | - | - |
Human Genome U133 Set | 208580_x_at | - | - |
Human Genome U133 Set | 232035_at | - | - |
Human Genome U133 Set | 234960_PM_at | - | - |
Human Genome U133 Set | 232035_PM_at | - | - |
Human Genome U133 Set | 234960_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031114 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031114 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 119151 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2119320 | NM_003540.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2119320 | BC069288.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777454 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_2119320 | NM_003540.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2119320 | BC069288.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2119320 | NM_003540.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2119320 | BC069288.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2119320 | NM_003540.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2119320 | BC069288.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21166391-S | NM_003540.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21166391-S | BC069288.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_29698-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0003610035 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0003610035 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1777454 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777454 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6860195 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_29698 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2119320 | NM_003540.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2119320 | BC069288.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2119320 | NM_003540.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2119320 | BC069288.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2119320 | NM_003540.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2119320 | BC069288.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1777454 | - | - |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2119320 | NM_003540.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2119320 | BC069288.1 | human_transcriptome |