Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERVFRD-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207582.2 | NP_997465.1 | 2 | 3254 | 117 | |
GenBank mRNA | AK075092.1 | - | 1 | 1620 | 117 | |
BC068585.1 | - | 2 | 3142 | 30346522 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15785245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552921 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15785262 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741242 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15785266 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741268 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741271 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552934 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552913 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552888 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741277 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741208 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552894 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741209 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552887 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741261 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552917 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741237 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15785247 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15785259 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741224 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552905 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552933 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741239 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741229 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741275 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741262 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552900 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741263 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552914 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552892 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04552908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741232 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741222 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741210 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741250 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741226 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741269 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24741236 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P924261 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P924261 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 44731 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 132 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 55666 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P924261 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P924261 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741266 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04552921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15785266 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741261 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04552921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741266 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15785266 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24741261 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P924261 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04552917 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04552922 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04552918 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04552914 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04552919 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04552916 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04552889 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15785252 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15785251 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15785255 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04552888 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15785254 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15785253 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01423009 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P924261 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 52031 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P31683459 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552912 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15785247 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552931 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552922 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552888 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552933 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552901 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552890 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552896 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552910 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15785266 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552892 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552916 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552925 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15785253 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552898 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552886 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15785250 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552919 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552928 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552908 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552927 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15785256 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552887 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552906 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552935 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552913 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552895 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04552914 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15785245 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_207582_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123931 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123933 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123932 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123934 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123930 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123935 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8123929 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1555880_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11755891_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11755891_a_at | - | - |
Affymetrix Human Genome U219 Array | 11725883_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11725883_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06015227.hg.1 | TC06001272.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06015227.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06015227.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06015227.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010807.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173501.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600173501.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600173501.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173501.hg.1 | ENST00000472091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600173501.hg.1 | ENST00000542862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600173501.hg.1 | NM_207582.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173501.hg.1 | TC0600010807.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000626665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007910.hg.1 | ENST00000627700.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1555880_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1555880_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_207582_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555880_at3 | - | - |
Human Genome U95 Set | 58940_at | - | - |
Human Genome U95 Set | 58940_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007053 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007053 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_21137-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6130484 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_21137 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1755392 | BC068585.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1755392 | BC068585.1 | human_transcriptome |