Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021918.hg.1 |
ENST00000239174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021918.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021918.hg.1 |
TC17001641.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021920.hg.1 |
ENST00000239174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021920.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021920.hg.1 |
TC17001641.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010990.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188392.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188392.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188392.hg.1 |
TC1700010990.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700188393.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188393.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188393.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188393.hg.1 |
ENST00000239174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188393.hg.1 |
ENST00000577092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188393.hg.1 |
NM_002144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188393.hg.1 |
TC1700010990.hg.1 |
Clariom D TC |