Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260861 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947770 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947754 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947776 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947767 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292930 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292903 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947781 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260866 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260868 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292924 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292887 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947759 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292905 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947748 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947750 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947760 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292907 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947753 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947779 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292920 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260862 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292911 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292929 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292888 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947773 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292885 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260867 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292918 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292883 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292904 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260871 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947755 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947758 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292923 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292908 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947768 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947756 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260870 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292899 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947747 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16260869 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P06947769 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27292927 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
AK126020.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
AK126229.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
AK312284.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
BC000534.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
BC007798.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
BC012799.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
BC013298.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
BC021203.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
CR541698.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
M26066.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
NM_000289.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
U24183.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
116635 |
Y00698.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
39146 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
62099 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3299639 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0359 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0360 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0361 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
17676 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P02174693 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P02174695 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3299639 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3299639 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292927 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292908 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292918 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292899 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292885 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292903 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292887 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947773 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292888 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260862 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947760 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260867 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947753 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947756 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947776 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947747 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292905 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260869 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292923 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292920 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947754 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947748 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947758 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947768 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260866 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947771 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947779 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947750 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947770 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260868 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260871 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947781 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260870 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292907 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292924 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947773 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947756 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260870 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947748 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292920 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292907 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947750 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947770 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260869 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292908 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947747 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292918 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292885 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260867 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947753 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260871 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947754 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292924 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260862 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947781 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947760 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292905 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947776 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292927 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947758 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260866 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947779 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292923 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16260868 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292887 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947771 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292888 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292899 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P27292903 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P06947768 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947776 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947759 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947749 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947771 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947768 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947753 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947783 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947775 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947758 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947767 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947757 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947773 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947762 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947778 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947756 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947755 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947766 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947748 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947747 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947764 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947765 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947752 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947770 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P06947781 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947745 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947742 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947776 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16260869 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947747 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16260866 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947764 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947748 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947785 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16260865 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947744 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947758 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947783 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16260868 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947762 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947768 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16260871 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947767 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947771 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947781 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947738 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947757 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947773 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947770 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947743 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16260867 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947778 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947750 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947754 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947766 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P06947775 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16260861 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P02174695 |
- |
- |