Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316364.1 | NP_001303293.1 | 3 - 4 | 533 | 151 | |
NM_003908.4 | NP_003899.2 | 3 - 4 | 533 | 151 | ||
XM_017028118.1 | XP_016883607.1 | 3 - 4 | 695 | 151 | ||
GenBank mRNA | AB451237.1 | - | 2 - 3 | 287 | 151 | |
AB451362.1 | - | 2 - 3 | 287 | 151 | ||
AK313025.1 | - | 3 - 4 | 404 | 151 | ||
BC000461.2 | - | 3 - 4 | 423 | 2822324 | 151 | |
BC000934.2 | - | 3 - 4 | 398 | 3447202 | 151 | |
BC007888.2 | - | 2 - 3 | 214 | 4120898 | 151 | |
BT009795.1 | - | 2 - 3 | 287 | 151 | ||
BX418731.2 | - | 3 - 4 | 403 | 151 | ||
CR456734.1 | - | 2 - 3 | 287 | 151 | ||
M29536.1 | - | 3 - 4 | 403 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |