Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609085 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152649 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152693 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609054 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609102 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152696 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152639 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152700 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609075 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609053 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152679 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609098 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152705 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152676 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152662 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609052 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152653 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152643 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152673 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609082 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609076 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609069 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152694 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609051 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152688 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609083 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609077 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609055 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609072 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152677 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152641 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16682787 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609110 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152654 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16682786 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152678 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609106 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609096 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609109 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609095 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152675 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152686 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609044 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609067 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609100 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609071 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609107 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609093 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609111 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609108 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609060 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609066 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152682 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609081 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152640 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09152659 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609094 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29609078 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3349751 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0666 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0667 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
22239 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P19918391 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3349751 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3349751 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609079 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16682790 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152651 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609083 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609095 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609051 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609100 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609060 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609085 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609071 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152693 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152679 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609055 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609075 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609068 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609067 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609069 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609098 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16682786 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609081 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609076 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609096 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152652 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609093 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609054 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609077 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609066 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16682787 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152676 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609078 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609052 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609094 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152682 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609085 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609100 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609083 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609096 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152652 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152682 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609079 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609093 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609075 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609051 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152693 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609052 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16682787 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609095 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609060 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609081 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609078 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609098 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609055 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609076 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609067 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609077 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16682786 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609066 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609094 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152651 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16682790 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609069 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152676 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609054 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09152679 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609071 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29609068 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P02870456 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152682 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152689 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152647 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152653 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152650 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152672 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152645 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152686 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152670 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152655 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152666 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152674 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152668 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152648 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152658 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152691 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152693 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152663 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152661 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152659 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152695 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09152679 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P02870456 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
103666 |
BC005219.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
103666 |
CR595772.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
103666 |
CR596134.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
103666 |
CR598435.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
103666 |
CR615916.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
103666 |
NM_002524.2 |
refseq_mrna_human_NM |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
103666 |
X02751.1 |
genbank_mrna_human |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P02870456 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152705 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152645 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152679 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152695 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152647 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16682794 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152696 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152701 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152726 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152724 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152661 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152727 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152668 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152682 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152720 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152650 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152733 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152735 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152672 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152656 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152722 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152700 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152648 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152659 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152698 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152719 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152729 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16682792 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152693 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152717 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152686 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152642 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152691 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152689 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152730 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152666 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152639 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16682791 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16682787 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152675 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152670 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152653 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152658 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09152640 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P02870456 |
- |
- |