Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003005.hg.1 |
ENST00000338965.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003005.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003005.hg.1 |
TC11000229.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003004.hg.1 |
ENST00000338965.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003004.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003004.hg.1 |
TC11000229.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003006.hg.1 |
ENST00000338965.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003006.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003006.hg.1 |
TC11000229.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100149029.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149029.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149029.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149029.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149029.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149029.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149029.hg.1 |
TC1100006966.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149027.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149027.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149027.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149027.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149027.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149027.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149027.hg.1 |
TC1100006966.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006966.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149030.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149030.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149030.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149030.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149030.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149030.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149030.hg.1 |
TC1100006966.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149026.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149026.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149026.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149026.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149026.hg.1 |
TC1100006966.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149028.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149028.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149028.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149028.hg.1 |
ENST00000338965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149028.hg.1 |
ENST00000530403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149028.hg.1 |
NM_001202439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149028.hg.1 |
TC1100006966.hg.1 |
Clariom D TC |