Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000561903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000564182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000565326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000568449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
ENST00000570069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006566.hg.1 |
NM_001257370.1 |
Refseq |