Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021242.hg.1 |
TC01001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021242.hg.1 |
ENST00000321935.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021242.hg.1 |
ENST00000339348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021242.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021242.hg.1 |
ENST00000392235.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021242.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175317.hg.1 |
TC0100010268.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000321935.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000339348.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000368106.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000392235.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000536257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000540741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
ENST00000541729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001004310.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010268.hg.1 |
NM_001284217.1 |
Refseq |