Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009614.2 | NP_001009614.1 | 2 | 275 | 114 | |
GenBank mRNA | DQ336118.1 | - | 2 | 275 | 114 | |
DQ336119.1 | - | 2 | 275 | 114 | ||
DQ336120.1 | - | 2 | 275 | 114 | ||
DQ336121.1 | - | 2 | 275 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739627 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004659 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739621 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739628 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004647 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004663 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004670 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739637 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004633 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739641 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739670 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739671 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739636 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739619 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739681 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739674 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004632 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739626 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004631 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739689 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739691 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739687 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004665 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739632 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12004673 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32739667 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 848678 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 844540 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 514026 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3276653 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0810 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0809 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57266 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3276653 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3276653 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739631 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739628 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004649 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004646 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17407801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17407792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004649 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739628 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17407801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004646 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17407792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12004640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32739631 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41829537 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004669 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004633 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004629 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004632 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004676 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004656 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004653 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004663 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004664 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004661 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004658 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004636 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004637 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004640 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004672 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004659 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004644 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004641 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004627 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004654 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004638 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004634 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004670 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12004675 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P27187060 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001009614_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170305 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170306 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170304 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8170302 | - | - |
Affymetrix Human Genome U219 Array | 11742652_x_at | - | - |
Affymetrix Human Genome U219 Array | 11742652_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008964.hg.1 | TC0X000700.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008964.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008962.hg.1 | TC0X000700.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008962.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008962.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008963.hg.1 | TC0X000700.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008963.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008963.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158570.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158570.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158570.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158570.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158570.hg.1 | TC0X00008638.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00158572.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158572.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158572.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158572.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158572.hg.1 | TC0X00008638.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00158571.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158571.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158571.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158571.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158571.hg.1 | TC0X00008638.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | ENST00000370493.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008638.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158573.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158573.hg.1 | NM_001009614.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158573.hg.1 | TC0X00008638.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041647 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041647 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_15038-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5670725 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_15038 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1654459 | DQ336121.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1654459 | NM_001009614.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1654459 | DQ336118.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1654459 | DQ336119.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1654459 | DQ336121.1 | human_transcriptome |