Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000507090.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006907.hg.1 |
ENST00000602906.1 |
Ensembl |