Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000302125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000417694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000503481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000509591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000511979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
ENST00000513389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012205.hg.1 |
NM_016459.3 |
Refseq |