Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058222.2 | NP_478129.1 | 2 | 208 | 70 | |
XM_017016604.1 | XP_016872093.1 | 2 | 202 | 70 | ||
GenBank mRNA | AF312827.1 | - | 2 | 129 | 70 | |
BC113497.1 | - | 2 | 208 | 8322549 | 70 | |
BC113499.1 | - | 2 | 208 | 8322551 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |