Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000106.5 | NP_000097.3 | 4 - 5 | 757 | 71 | |
NM_001025161.2 | NP_001020332.2 | 3 - 4 | 604 | 71 | ||
XM_011529966.2 | XP_011528268.1 | 4 - 5 | 751 | 71 | ||
XM_011529968.2 | XP_011528270.1 | 4 - 5 | 751 | 71 | ||
XM_011529970.2 | XP_011528272.1 | 3 - 4 | 598 | 71 | ||
XM_011529972.2 | XP_011528274.1 | 4 - 5 | 751 | 71 | ||
GenBank mRNA | AK309600.1 | - | 3 - 4 | 602 | 71 | |
AY663390.1 | - | 2 - 3 | 338 | 71 | ||
BC066877.1 | - | 3 - 4 | 515 | 71 | ||
BC067432.1 | - | 4 - 5 | 668 | 7002157 | 71 | |
BC075023.2 | - | 4 - 5 | 697 | 30915411 | 71 | |
BC075024.2 | - | 4 - 5 | 697 | 30915622 | 71 | |
BC106757.1 | - | 3 - 4 | 536 | 40024713 | 71 | |
BC106758.1 | - | 4 - 5 | 689 | 40024717 | 71 | |
BC126858.1 | - | 4 - 5 | 689 | 40024719 | 71 | |
BX422592.2 | - | 4 - 5 | 742 | 71 | ||
CR456430.1 | - | 3 - 4 | 539 | 71 | ||
M20403.1 | - | 4 - 5 | 667 | 71 | ||
X07618.1 | - | 3 - 4 | 369 | 71 | ||
X08006.1 | - | 4 - 5 | 667 | 71 |
Gene Symbol: LOC101929829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111920.1 | 4 - 5 | 756 | 71 | ||
GenBank mRNA | AB209492.1 | - | 3 - 4 | 2935 | 71 | |
AK308211.1 | - | 4 - 5 | 751 | 71 | ||
M24499.1 | - | 2 - 3 | 295 | 71 | ||
X16865.1 | - | 2 - 3 | 301 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection P450 Detect Genomic DNA Homo sapiens Inventoried Probe spans exons |