Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316363.1 | NP_001303292.1 | 4 - 5 | 669 | 149 | |
NM_001316364.1 | NP_001303293.1 | 4 - 5 | 678 | 149 | ||
NM_003908.4 | NP_003899.2 | 4 - 5 | 678 | 149 | ||
XM_017028118.1 | XP_016883607.1 | 4 - 5 | 840 | 149 | ||
GenBank mRNA | AB451237.1 | - | 3 - 4 | 432 | 149 | |
AB451362.1 | - | 3 - 4 | 432 | 149 | ||
AK313025.1 | - | 4 - 5 | 549 | 149 | ||
BC000461.2 | - | 4 - 5 | 568 | 2822324 | 149 | |
BC000934.2 | - | 4 - 5 | 543 | 3447202 | 149 | |
BC007888.2 | - | 3 - 4 | 359 | 4120898 | 149 | |
BT009795.1 | - | 3 - 4 | 432 | 149 | ||
BX418731.2 | - | 4 - 5 | 548 | 149 | ||
BX429051.2 | - | 4 - 5 | 515 | 149 | ||
CR456734.1 | - | 3 - 4 | 432 | 149 | ||
M29536.1 | - | 4 - 5 | 548 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |