Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198508.2 | NP_940910.1 | 2 - 3 | 925 | 104 | |
XM_005250311.3 | XP_005250368.1 | 2 - 3 | 930 | 104 | ||
XM_011516140.2 | XP_011514442.1 | 2 - 3 | 930 | 104 | ||
XM_011516141.2 | XP_011514443.1 | 2 - 3 | 930 | 104 | ||
GenBank mRNA | AK126174.1 | - | 2 - 3 | 930 | 104 | |
BC104963.1 | - | 2 - 3 | 890 | 8143966 | 104 | |
BC104965.1 | - | 2 - 3 | 890 | 8143968 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons |