Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000339686.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000392206.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000392207.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000402589.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000414866.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000418261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000419602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000424570.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000429837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000436012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000440396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000443640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000447113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000451297.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000458071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
ENST00000462721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_001126056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_001126057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_001126058.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_001126059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_001190347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_001190348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_001190349.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NM_033317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
NR_033746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019491.hg.1 |
TC19001445.hg.1 |
HTA TC |