Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020179.hg.1 |
TC06001692.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020179.hg.1 |
ENST00000373365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020179.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020178.hg.1 |
TC06001692.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020178.hg.1 |
ENST00000373365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020178.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020183.hg.1 |
TC06001692.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020183.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020177.hg.1 |
TC06001692.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020177.hg.1 |
ENST00000373365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020177.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180447.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180447.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180447.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180447.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180447.hg.1 |
TC0600011725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600180448.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180448.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180448.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180448.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180448.hg.1 |
TC0600011725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600180446.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180446.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180446.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180446.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180446.hg.1 |
TC0600011725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
ENST00000470973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011725.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180449.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180449.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180449.hg.1 |
ENST00000373365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180449.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180449.hg.1 |
TC0600011725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600180453.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180453.hg.1 |
NM_006708.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180453.hg.1 |
TC0600011725.hg.1 |
Clariom D TC |