Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014180.hg.1 |
ENST00000438700.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014180.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014180.hg.1 |
TC10001132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169035.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169035.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169035.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169035.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169035.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169035.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169035.hg.1 |
TC1000010129.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000438700.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
ENST00000622555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010129.hg.1 |
NM_001034842.3 |
Refseq |