Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003834.hg.1 |
TC01000242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003834.hg.1 |
ENST00000400664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003834.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007144.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149789.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149789.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149789.hg.1 |
ENST00000400664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149789.hg.1 |
NM_152375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149789.hg.1 |
TC0100007144.hg.1 |
Clariom D TC |