Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_014566_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8011321 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8003807 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8011320 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8003806 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006574.hg.1 |
ENST00000525719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006574.hg.1 |
ENST00000525719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006574.hg.1 |
ENST00000531680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006574.hg.1 |
NR_033795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013350.hg.1 |
NM_014566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013350.hg.1 |
TC17001001.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009469.hg.1 |
ENST00000575751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009469.hg.1 |
NM_014566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172415.hg.1 |
ENST00000575751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172415.hg.1 |
NM_014566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172415.hg.1 |
ENST00000575751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172415.hg.1 |
NM_014566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172415.hg.1 |
TC1700009469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006574.hg.1 |
ENST00000525719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006574.hg.1 |
ENST00000525719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006574.hg.1 |
ENST00000531680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006574.hg.1 |
NR_033795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009469.hg.1 |
ENST00000575751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009469.hg.1 |
NM_014566.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
221341_PM_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
221341_s_at |
- |
- |
Human Genome U133A 2.0 Array |
221341_PM_s_at |
- |
- |
Human Genome U133A 2.0 Array |
221341_s_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_014566_at |
- |
- |
Human Genome U133 Set |
221341_PM_s_at |
- |
- |
Human Genome U133 Set |
221341_s_at |
- |
- |