Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_033059_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015222 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019700 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007117 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019573 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007116 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019572 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007114 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019571 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015231 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015219 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019567 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007113 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015227 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8019570 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015221 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015230 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8019699 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8007112 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8007115 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
234633_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11745692_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11745692_PM_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019718.hg.1 |
ENST00000391413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019718.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019718.hg.1 |
TC17001489.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019717.hg.1 |
ENST00000391413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019717.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019717.hg.1 |
TC17001489.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206387.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206387.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206387.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206387.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206387.hg.1 |
TC1700012406.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206388.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206388.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206388.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206388.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206388.hg.1 |
TC1700012406.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206389.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206389.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206389.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206389.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206389.hg.1 |
TC1700012406.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Human Genome U133 Plus 2.0 Array |
234633_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
234633_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234633_at10 |
- |
- |
Human Genome U133 Set |
234633_at |
- |
- |
Human Genome U133 Set |
234633_PM_at |
- |
- |