Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM24B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204364.1 | NP_001191293.1 | 4 | 534 | 110 | |
NM_152644.2 | NP_689857.2 | 4 | 564 | 110 | ||
NR_037911.1 | 3 | 437 | 110 | |||
GenBank mRNA | AL832386.1 | - | 1 | 1520 | 110 | |
AL832795.1 | - | 1 | 1666 | 110 | ||
BC031343.1 | - | 4 | 485 | 5456151 | 110 | |
BG490871.1 | - | 4 | 499 | 110 | ||
BU602099.1 | - | 4 | 447 | 110 | ||
BU958134.1 | - | 3 | 351 | 110 | ||
CB410280.1 | - | 3 | 438 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon |