Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000417645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000417645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011724.hg.1 |
TC02000752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011724.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011724.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011724.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011724.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163481.hg.1 |
TC0200009067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000306406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000409826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000417645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000417645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
ENST00000465296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009067.hg.1 |
NM_183240.2 |
Refseq |