Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000498210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000498210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
ENST00000498210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000391621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000498210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000498210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
ENST00000498210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NM_198693.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008376.hg.1 |
NR_130165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149937.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149937.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149937.hg.1 |
TC2100007376.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100149937.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149937.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149938.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149938.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149938.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149938.hg.1 |
TC2100007376.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100149938.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149938.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149938.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149941.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149941.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149941.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149941.hg.1 |
TC2100007376.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100149941.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149941.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149941.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000397911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000484861.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
ENST00000616529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007376.hg.1 |
NM_198690.2 |
Refseq |