Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007797.hg.1 |
ENST00000403458.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007797.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007797.hg.1 |
TC16000545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007799.hg.1 |
ENST00000403458.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007799.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007799.hg.1 |
TC16000545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007801.hg.1 |
ENST00000403458.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007801.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007801.hg.1 |
TC16000545.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160920.hg.1 |
TC1600008170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160921.hg.1 |
TC1600008170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008170.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160919.hg.1 |
TC1600008170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000602974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160926.hg.1 |
TC1600008170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000403458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000445068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000459925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000602365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
ENST00000602987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
NM_001012984.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600160924.hg.1 |
TC1600008170.hg.1 |
Clariom D TC |