Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008058.hg.1 |
ENST00000216927.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008058.hg.1 |
ENST00000303415.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008058.hg.1 |
ENST00000381580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008058.hg.1 |
ENST00000381583.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008058.hg.1 |
NM_001039508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008058.hg.1 |
NM_018556.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008058.hg.1 |
TC20000555.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
ENST00000216927.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
ENST00000303415.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
ENST00000344103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
ENST00000381580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
ENST00000381583.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
ENST00000478145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
NM_001039508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
NM_018556.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
NM_080816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008054.hg.1 |
TC20000555.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000216927.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000303415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000381580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000381583.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
NM_001039508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
NM_018556.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
TC2000009971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000216927.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000303415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000381580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
ENST00000381583.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
NM_001039508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170927.hg.1 |
NM_018556.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000216927.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000303415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000344103.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000381580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000381583.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000478145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000497407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
NM_001039508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
NM_018556.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
NM_080816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
TC2000009971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000216927.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000303415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000344103.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000381580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000381583.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000478145.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
ENST00000497407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
NM_001039508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
NM_018556.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170923.hg.1 |
NM_080816.2 |
Refseq |