Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019058.2 | NP_061931.1 | 2 - 3 | 411 | 90 | |
GenBank mRNA | AF335324.1 | - | 2 - 3 | 427 | 90 | |
AK000507.1 | - | 2 - 3 | 406 | 90 | ||
AL136668.1 | - | 2 - 3 | 410 | 90 | ||
AY090097.1 | - | 2 - 3 | 327 | 90 | ||
BC000708.1 | - | 2 - 3 | 317 | 3507244 | 90 | |
BC007714.1 | - | 2 - 3 | 399 | 4302878 | 90 | |
BC015236.1 | - | 2 - 3 | 385 | 3906374 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons |