Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080725.2 | NP_542763.1 | 1 - 2 | 396 | 87 | |
GenBank mRNA | AK127435.1 | - | 1 - 2 | 241 | 87 | |
AK312054.1 | - | 1 - 2 | 226 | 87 | ||
AL833944.1 | - | 1 - 2 | 272 | 87 | ||
BC017001.1 | - | 1 - 2 | 191 | 87 | ||
BC032604.1 | - | 1 - 2 | 227 | 5535883 | 87 | |
BC047707.1 | - | 1 - 2 | 216 | 5735551 | 87 | |
BU170568.1 | - | 361 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons |