Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010446.hg.1 |
TC06000883.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010446.hg.1 |
ENST00000368876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010446.hg.1 |
ENST00000368877.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010446.hg.1 |
ENST00000368882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010446.hg.1 |
ENST00000368885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010446.hg.1 |
ENST00000451850.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010446.hg.1 |
NM_001634.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368876.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368885.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000451850.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000610605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000612642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000619590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001287214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001287215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001287216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001634.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NR_109768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368876.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000368885.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000451850.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000610605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000612642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
ENST00000619590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001287214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001287215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001287216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NM_001634.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
NR_109768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162690.hg.1 |
TC0600009127.hg.1 |
Clariom D TC |