Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040655.hg.1 |
TC01002611.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040655.hg.1 |
ENST00000262746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040655.hg.1 |
ENST00000319248.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040655.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040655.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040655.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040655.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000372079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
ENST00000483583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014014.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000262746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000319248.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000424390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
ENST00000447184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_001202431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_002574.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_181696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
NM_181697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100204805.hg.1 |
TC0100014014.hg.1 |
Clariom D TC |