Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY740634.1 | - | 11 | 933 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Inventoried Amplicon spans exons and probe does not span exons |