Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HOXA11 antisense RNA | HOXA11-AS |
HOXA-AS5, HOXA11-AS1, HOXA11AS, HOXA11S, NCRNA00076 |
-1.7kb | Sense | Chr. 7 - 27185408 - 27189293 on NCBI Build GRCh38 |
Downstream | homeobox A13 | HOXA13 |
HOX1, HOX1J |
2.0kb | Anti-sense | Chr. 7 - 27196880 - 27200106 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |