Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105379655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK097625.1 | - | 6 | 1364 | 151 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
major histocompatibility complex, class I, H (pseudogene) |
HLA-H | HLAHP | -8.3kb | Anti-sense | Chr. 6 - 29887760 - 29891080 on NCBI Build GRCh38 |
Downstream | HLA complex group 4B (non-protein coding) | HCG4B |
HCG4P6, HCGIV-06, HCGIV-6, HCGIV.5, bCX67J3.3, bPG309N1.1, bQB90C11.3 |
23.6kb | Sense | Chr. 6 - 29924592 - 29927215 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |