Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSX2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002307.1 | 1 | 1350 | 114 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P169179 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P169179 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 30263 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7292 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P169179 | - | - |
SurePrint G3 Human GE Microarray G4851A | ERCC-00028_121 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P169179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392474 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30970801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392481 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30970812 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392476 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392483 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30970799 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392483 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30970801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392476 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392474 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392481 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30970812 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30970799 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10392485 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P169179 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P169179 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16999948 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392527 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392441 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392439 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392436 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392509 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392518 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392501 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392427 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392532 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16999963 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392459 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392434 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392449 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392451 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392516 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392474 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392475 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392447 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392504 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392418 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392428 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392470 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392482 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392495 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392503 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16999944 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392523 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392472 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392511 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392493 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392506 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392528 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392486 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392521 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392421 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392462 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392425 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392455 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392526 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16999946 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392445 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392443 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392438 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10392499 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_002307_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8008717 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8008716 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR17009250.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17009250.hg.1 | TC17000709.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162632.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162632.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162632.hg.1 | TC1700008422.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008422.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162628.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162628.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162628.hg.1 | TC1700008422.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700162630.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700162630.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162630.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700162630.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162630.hg.1 | TC1700008422.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700162629.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700162629.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162629.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700162629.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162629.hg.1 | TC1700008422.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700162631.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700162631.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162631.hg.1 | ENST00000584100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700162631.hg.1 | NR_002307.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700162631.hg.1 | TC1700008422.hg.1 | Clariom D TC |
Human Genome U95 Set | 702_f_at | - | - |
Human Genome U95 Set | 702_PM_f_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009770 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009769 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009769 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009770 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 228070 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2197647 | NR_002307.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1688723 | NR_002307.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1688723 | NR_002307.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2197647 | NR_002307.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1688723 | NR_002307.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2197647 | NR_002307.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2197647 | NR_002307.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1688723 | NR_002307.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_20810 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_20810 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2197647 | NR_002307.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2197647 | NR_002307.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1688723 | NR_002307.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1688723 | NR_002307.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2197647 | NR_002307.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1688723 | NR_002307.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2197647 | NR_002307.1 | human_transcriptome |