Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043533.hg.1 |
TC02002701.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043533.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043533.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043534.hg.1 |
TC02002701.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043534.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043534.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043535.hg.1 |
TC02002701.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043535.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043535.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213988.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213988.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213988.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213988.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213988.hg.1 |
TC0200015525.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200213989.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213989.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213989.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213989.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213989.hg.1 |
TC0200015525.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200213991.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213991.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213991.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213991.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213991.hg.1 |
TC0200015525.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |