Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000495760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013169.hg.1 |
ENST00000495760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000247992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000429261.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000495760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013169.hg.1 |
ENST00000495760.2 |
Ensembl |