Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR11H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005239.1 | NP_001005239.1 | 1 | 750 | 160 |
Gene Symbol: OR11H12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013354.1 | NP_001013372.1 | 1 | 750 | 160 | |
GenBank mRNA | BC136730.1 | - | 1 | 822 | 9020720 | 160 |
BC136731.1 | - | 1 | 822 | 9020721 | 160 |
Gene Symbol: OR11H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197287.1 | NP_001184216.1 | 2 | 822 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CNV Microarray 2x400K | 5025 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P128868 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0592 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0593 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P128868 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 11432 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29237 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P128868 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P128868 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P128868 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P128868 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29604404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29604404 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P128868 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P128868 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P418230 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7977473 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7977451 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8074195 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7977473 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972982 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7977451 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8074195 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972982 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7972981 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7977450 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8074194 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7977472 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7977450 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8074194 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7972981 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7977472 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR14000013.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14000013.hg.1 | NM_001013354.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14000013.hg.1 | TC14000006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22008330.hg.1 | ENST00000252835.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22008330.hg.1 | NM_001005239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22008330.hg.1 | TC22000458.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14009505.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009505.hg.1 | TC14000871.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500008675.hg.1 | ENST00000554547.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006450.hg.1 | ENST00000252835.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006450.hg.1 | NM_001005239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006466.hg.1 | ENST00000359695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006446.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006446.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006446.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006446.hg.1 | NM_001013354.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009506.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009506.hg.1 | TC14000871.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500008663.hg.1 | ENST00000624197.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008568.hg.1 | ENST00000556246.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008568.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008568.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008568.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008568.hg.1 | ENST00000556246.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008568.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008568.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008568.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008675.hg.1 | ENST00000554547.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200144073.hg.1 | TC2200006450.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200144073.hg.1 | ENST00000252835.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200144073.hg.1 | NM_001005239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200144073.hg.1 | ENST00000252835.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200144073.hg.1 | NM_001005239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400144071.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144071.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144071.hg.1 | TC1400006446.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400161489.hg.1 | ENST00000556246.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400161489.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400161489.hg.1 | ENST00000556246.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400161489.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400161489.hg.1 | TC1400008568.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400161488.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400161488.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400161488.hg.1 | TC1400008568.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400161491.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400161491.hg.1 | NM_001197287.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400161491.hg.1 | TC1400008568.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC2200006450.hg.1 | ENST00000252835.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006450.hg.1 | NM_001005239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400144070.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144070.hg.1 | NM_001013354.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400144070.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144070.hg.1 | NM_001013354.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400144070.hg.1 | TC1400006446.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500008663.hg.1 | ENST00000624197.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006466.hg.1 | ENST00000359695.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144069.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144069.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144069.hg.1 | TC1400006446.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400006446.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006446.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006446.hg.1 | ENST00000550708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006446.hg.1 | NM_001013354.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051219 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019412 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019412 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019412 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019412 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019412 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019412 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051219 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2342651 | BC136731.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2342651 | BC136730.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1679870 | BC136731.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2342651 | BC136731.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2342651 | BC136730.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2342651 | BC136731.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2342651 | BC136730.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2342651 | BC136731.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2342651 | BC136730.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_13826-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 2510259 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13826 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2342651 | BC136731.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2342651 | BC136730.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2342651 | BC136731.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2342651 | BC136730.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2342651 | BC136731.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2342651 | BC136730.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1679870 | BC136730.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1669995 | NM_001005239.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1669995 | NM_001197287.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2072401 | NM_001005239.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2072401 | NM_001197287.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2342651 | BC136731.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2342651 | NM_001013354.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2342651 | BC136730.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1679870 | BC136731.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1679870 | NM_001013354.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1679870 | BC136730.1 | human_transcriptome |