Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 3 member B | FAM3B |
2-21, C21orf11, C21orf76, ORF9, PANDER, PRED44 |
Sense | Chr. 21 - 41304229 - 41357727 on NCBI Build GRCh38 | |
Downstream | MX dynamin like GTPase 2 | MX2 | MXB | 45.4kb | Sense | Chr. 21 - 41361986 - 41408943 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |