Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004565.hg.1 |
TC09000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004565.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004564.hg.1 |
TC09000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004564.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004563.hg.1 |
TC09000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004563.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007945.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153442.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153442.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153442.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153442.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153442.hg.1 |
TC0900007945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153449.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153449.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153449.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153449.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153449.hg.1 |
TC0900007945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153444.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153444.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153444.hg.1 |
ENST00000412768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153444.hg.1 |
NR_015444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153444.hg.1 |
TC0900007945.hg.1 |
Clariom D TC |