Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000441.hg.1 |
ENST00000320936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000441.hg.1 |
ENST00000444172.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000441.hg.1 |
NM_001280.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000441.hg.1 |
NR_023312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000441.hg.1 |
NR_023313.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000441.hg.1 |
TC19000034.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000320936.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000585630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000585913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000585914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586636.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000587169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000587323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000587896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000589235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000589686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000589710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000590171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000590704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000592412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000628979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NM_001280.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NM_001300815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NM_001300829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NR_023312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NR_023313.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
TC1900006524.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000320936.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000585630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000585913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000585914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586636.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000586773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000587169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000587323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000587896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000588411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000589235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000589686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000589710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000590171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000590704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000591935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000592412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000593283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
ENST00000628979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NM_001280.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NM_001300815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NM_001300829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NR_023312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145108.hg.1 |
NR_023313.2 |
Refseq |