Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003827.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003827.hg.1 |
TC17000318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152266.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152266.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152266.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152266.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152266.hg.1 |
TC1700007357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700152267.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152267.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152267.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152267.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152267.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152267.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152267.hg.1 |
TC1700007357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000225992.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000587926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591228.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
ENST00000591703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010806.hg.1 |
NM_002722.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152265.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152265.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152265.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152265.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152265.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152265.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152265.hg.1 |
TC1700007357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000582808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
ENST00000583761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007357.hg.1 |
NR_002181.1 |
Refseq |