Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006645.hg.1 |
TC0X002320.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006645.hg.1 |
ENST00000243286.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006645.hg.1 |
ENST00000372627.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006645.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006645.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006644.hg.1 |
TC0X002320.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006644.hg.1 |
ENST00000243286.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006644.hg.1 |
ENST00000372627.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006644.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006644.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177835.hg.1 |
TC0X00011315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000372774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
ENST00000622517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010380.hg.1 |
NM_001006938.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000243286.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
ENST00000372628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_001006933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011315.hg.1 |
NM_032926.2 |
Refseq |