Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100129775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK094853.1 | - | 1 | 2153 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microtubule associated protein 6 | MAP6 |
MAP6-N, MTAP6, N-STOP, STOP |
Sense | Chr. 11 - 75586914 - 75671108 on NCBI Build GRCh38 | |
Downstream | monoacylglycerol O-acyltransferase 2 | MOGAT2 |
DGAT2L5, DGAT2L5., MGAT2, hDC5 |
115.8kb | Anti-sense | Chr. 11 - 75717819 - 75732958 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |